Please login to your online account to display your discounted pricing

5-Bromoindole-3-acetic Acid 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107072500

 View more versions of this product

Catalog No. AC107072500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 254.08
Quantity 250mg
Color Off-white to beige
EINECS Number 254-917-8
Melting Point 141° to 148°C
CAS Max % 100.0
Water 1% max. (K.F.)
Assay Percent Range 99%
Infrared Spectrum Authentic
Chemical Name or Material 5-Bromoindole-3-acetic acid
Packaging Glass bottle
Name Note 99%
Molecular Formula C10H8BrNO2
MDL Number MFCD00005637
CAS 40432-84-6
CAS Min % 98.5
Health Hazard 1 LIGHT SENSITIVE

Specifications

Chemical Name or Material 5-Bromoindole-3-acetic acid
Name Note 99%
CAS 40432-84-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8BrNO2
Formula Weight 254.08
MDL Number MFCD00005637
Accessories

LIGHT SENSITIVE

SDS & Safety Documents