missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromoindole-3-acetic acid, 99%, ACROS Organics™

Chemical Identifiers

CAS 40432-84-6
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.08
MDL Number MFCD00005637
InChI Key WTFGHMZUJMRWBK-UHFFFAOYSA-N
Synonym 1h-indole-3-acetic acid, 5-bromo, 2-5-bromo-1h-indol-3-yl acetic acid, 4ojq, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 5-bromo-1h-indole-3-acetic acid, 5-bromo-3-indoleacetic acid, 5-bromoindole-3-acetic acid, 5-bromoindole-3-aceticacid, chembl82440
PubChem CID 96734
IUPAC Name 2-(5-bromo-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC107072500
SDS View Documents
Thermo Fisher Scientific
107072500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC107070010
SDS View Documents
Thermo Fisher Scientific
107070010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

40432-84-6
254.08
WTFGHMZUJMRWBK-UHFFFAOYSA-N
96734
C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O
C10H8BrNO2
MFCD00005637
1h-indole-3-acetic acid, 5-bromo, 2-5-bromo-1h-indol-3-yl acetic acid, 4ojq, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 5-bromo-1h-indole-3-acetic acid, 5-bromo-3-indoleacetic acid, 5-bromoindole-3-acetic acid, 5-bromoindole-3-aceticacid, chembl82440
2-(5-bromo-1H-indol-3-yl)acetic acid

Specifications

Authentic
1% max. (K.F.)
141°C to 148°C
40432-84-6
100.0
C10H8BrNO2
1h-indole-3-acetic acid, 5-bromo, 2-5-bromo-1h-indol-3-yl acetic acid, 4ojq, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 5-bromo-1h-indole-3-acetic acid, 5-bromo-3-indoleacetic acid, 5-bromoindole-3-acetic acid, 5-bromoindole-3-aceticacid, chembl82440
WTFGHMZUJMRWBK-UHFFFAOYSA-N
2-(5-bromo-1H-indol-3-yl)acetic acid
96734
Crystalline Powder
5-Bromoindole-3-acetic acid, 99%
Glass bottle
White to Beige
250mg
98.5
99%
MFCD00005637
Solubility in water: insoluble in water
C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O
254.08
254.08
99%
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 254-917-8

SDS
Product Certifications