missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromocytosine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 165782500

 View more versions of this product

Catalog No. AC165782500



Specifications

Specifications

5-Bromocytosine
Glass bottle
240.0°C to 243.0°C
99%
98.5
99%
MFCD00056025
2 1h-pyrimidinone, 4-amino-5-bromo
C1=NC(=O)NC(=C1Br)N
190
190
99%
Authentic
Yellow
250mg
2240-25-7
100.0
C4H4BrN3O
24, 319
QFVKLKDEXOWFSL-UHFFFAOYSA-N
6-amino-5-bromo-1H-pyrimidin-2-one
75233
Amorphous Powder

Chemical Identifiers

2240-25-7
190
QFVKLKDEXOWFSL-UHFFFAOYSA-N
75233
C1=NC(=O)NC(=C1Br)N
C4H4BrN3O
MFCD00056025
2 1h-pyrimidinone, 4-amino-5-bromo
6-amino-5-bromo-1H-pyrimidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
If skin irritation occurs: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 218-806-8

SDS Documents