Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromobenzofurazan, 97%

$73.43 - $73.43

Chemical Identifiers

Chemical Name or Material 5-Bromobenzofurazan
CAS 51376-06-8
Molecular Formula C6H3BrN2O
Formula Weight 199.01
MDL Number MFCD02682026
Synonym 5-Bromo-2,1,3-benzoxadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5167403

  SDS
alfa aesar™
H5167403
1g Each for $73.43
Description & Specifications

Specifications

Formula Weight 199.01
Chemical Name or Material 5-Bromobenzofurazan
Quantity 1g
Assay 97%
Molecular Formula C6H3BrN2O
MDL Number MFCD02682026
CAS 51376-06-8
Synonym 5-Bromo-2,1,3-benzoxadiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.