Please login to your online account to display your discounted pricing

5-Bromobenzo[b]thiophene-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrO2S
Formula Weight 257.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC33501CB

  SDS
fisher scientific company
CC33501CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC33501DE

 
fisher scientific company
CC33501DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC33501DA

  SDS
fisher scientific company
CC33501DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.11
Quantity 250mg
Assay 97%
Molecular Formula C9H5BrO2S
CAS 7312-24-5
Packaging Amber Glass Bottle
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carboxylic acid
Assay Percent Range 97%