Please login to your online account to display your discounted pricing

5-Bromobenzo[b]thiophene-3-carboxylic acid, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC33501DE

 View more versions of this product

Catalog No. CC33501DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 257.11
Quantity 5g
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrO2S
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carboxylic acid
CAS 7312-24-5
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrO2S
Formula Weight 257.11