missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromobenzo[b]thiophene-3-carbaldehyde, 97%, Maybridge

$325.38 - $325.38

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H5BrOS
Formula Weight 241.11
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC33504DE Fisher Scientific Company
CC33504DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33504DA SDS Fisher Scientific Uk Ltd
CC33504DA
Amber Glass Bottle 1g Each for $325.38
CC33504CB SDS Fisher Scientific Company
CC33504CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
241.11
97%
C9H5BrOS
5-Bromobenzo[b]thiophene-3-carbaldehyde

Specifications

16296-72-3
5g
C9H5BrOS
5-Bromobenzo[b]thiophene-3-carbaldehyde
Amber Glass Bottle
97%
241.11
97%