Please login to your online account to display your discounted pricing

5-Bromobenzo[b]thiophene-3-carbaldehyde, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC33504CB

 View more versions of this product

Catalog No. CC33504CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 241.11
Quantity 250mg
Assay 97%
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carbaldehyde
Molecular Formula C9H5BrOS
Assay Percent Range 97%
CAS 16296-72-3

Specifications

Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrOS
Formula Weight 241.11