Please login to your online account to display your discounted pricing

(5-Bromobenzo[b]furan-2-yl)methylamine, ≥97%, Maybridge

$136.18 - $136.18

Chemical Identifiers

Chemical Name or Material (5-Bromobenzo[b]furan-2-yl)methylamine
CAS 165736-51-6
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H8BrNO
Formula Weight 226.07
MDL Number MFCD06658983
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC31213CB

  SDS
fisher scientific company
CC31213CB
Each for $136.18
Description & Specifications

Specifications

Formula Weight 226.07
Quantity 250mg
Assay 95%
Assay Percent Range 95%
Chemical Name or Material (5-Bromobenzo[b]furan-2-yl)methylamine
Molecular Formula C9H8BrNO
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
MDL Number MFCD06658983
CAS 165736-51-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.