Please login to your online account to display your discounted pricing

(5-Bromobenzo[b]furan-2-yl)methylamine, ≥97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31213DA

 View more versions of this product

Catalog No. CC31213DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.07
Quantity 1g
Assay 95%
CAS 165736-51-6
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (5-Bromobenzo[b]furan-2-yl)methylamine
MDL Number MFCD06658983
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Assay Percent Range 95%
Molecular Formula C9H8BrNO

Specifications

Chemical Name or Material (5-Bromobenzo[b]furan-2-yl)methylamine
CAS 165736-51-6
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H8BrNO
Formula Weight 226.07
MDL Number MFCD06658983
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

SDS & Safety Documents