Please login to your online account to display your discounted pricing

5-Bromobenzo[b]furan-2-carboxylic acid 97+%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31201DA

 View more versions of this product

Catalog No. CC31201DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 241.04
Quantity 1g
Assay 97%
Chemical Name or Material 5-Bromo-1-benzofuran-2-carboxylic acid
Assay Percent Range 97%
Molecular Formula C9H5BrO3
CAS 10242-11-2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 5-Bromo-1-benzofuran-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrO3
Formula Weight 241.04