Please login to your online account to display your discounted pricing

5-Bromobenzo[b]furan-2-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31204DA

 View more versions of this product

Catalog No. CC31204DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 225.04
Quantity 1g
Assay 97%
Chemical Name or Material 5-Bromo-1-benzofuran-2-carbaldehyde
Assay Percent Range 97%
Molecular Formula C9H5BrO2
CAS 23145-16-6
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 5-Bromo-1-benzofuran-2-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5BrO2
Formula Weight 225.04