Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromoacetyl-2-hydroxybenzamide, 95%

$69.27 - $204.73

Chemical Identifiers

Chemical Name or Material 5-Bromoacetyl-2-hydroxybenzamide
CAS 73866-23-6
Molecular Formula C9H8BrNO3
Formula Weight 258.07
MDL Number MFCD00792456
Synonym 5-(Bromoacetyl)salicylamide; 2-Bromo-3'-carboxamido-4'-hydroxyacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2433018

  SDS
alfa aesar™
B2433018
50g Each for $69.27

AAB2433030

  SDS
alfa aesar™
B2433030
250g Each for $204.73
Description & Specifications

Specifications

Formula Weight 258.07
Chemical Name or Material 5-Bromoacetyl-2-hydroxybenzamide
Assay 95%
Molecular Formula C9H8BrNO3
MDL Number MFCD00792456
CAS 73866-23-6
Melting Point 202° to 204°C
Synonym 5-(Bromoacetyl)salicylamide; 2-Bromo-3'-carboxamido-4'-hydroxyacetophenone
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CV2365900
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.