missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-N,N-dimethylpyridine-2-carboxamide, 96%

Manufacturer:  Alfa Aesar™ H5468603

 View more versions of this product

Catalog No. AAH5468603



Description

Description

Applications
5-Bromo-N,N-dimethylpyridine-2-carboxamide is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

5-Bromo-N,N-dimethylpyridine-2-carboxamide
845305-86-4
MFCD07357419
KQRHFAPOFNDZHY-UHFFFAOYSA-N
5-bromo-N,N-dimethylpyridine-2-carboxamide
22831618
96%
1g
C8H9BrN2O
5-bromo-n,n-dimethyl-pyridine-2-carboxamide
CN(C)C(=O)C1=NC=C(C=C1)Br
229.077
229.07

Chemical Identifiers

845305-86-4
229.077
KQRHFAPOFNDZHY-UHFFFAOYSA-N
22831618
CN(C)C(=O)C1=NC=C(C=C1)Br
C8H9BrN2O
MFCD07357419
5-bromo-n,n-dimethyl-pyridine-2-carboxamide
5-bromo-N,N-dimethylpyridine-2-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents