missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-N-methylpyridine-2-carboxamide, 96%

Manufacturer:  Alfa Aesar™ H5494703

 View more versions of this product

Catalog No. AAH5494703



Specifications

Specifications

5-Bromo-N-methylpyridine-2-carboxamide
1g
C7H7BrN2O
2-pyridinecarboxamide, 5-bromo-n-methyl
CNC(=O)C1=NC=C(C=C1)Br
215.05
215.05
76°C to 77°C
845305-87-5
MFCD07357421
OHYCXPXWHPWYFX-UHFFFAOYSA-N
5-bromo-N-methylpyridine-2-carboxamide
22831620
96%

Chemical Identifiers

845305-87-5
215.05
OHYCXPXWHPWYFX-UHFFFAOYSA-N
22831620
CNC(=O)C1=NC=C(C=C1)Br
C7H7BrN2O
MFCD07357421
2-pyridinecarboxamide, 5-bromo-n-methyl
5-bromo-N-methylpyridine-2-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents