Learn More
5-Bromo-m-xylene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 294000250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Bromo-m-xylene | |
1.3600g/mL | |
Authentic | |
Glass bottle | |
(CH3)2C6H3Br | |
25g | |
5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene | |
CC1=CC(=CC(=C1)Br)C | |
185.06 | |
185.06 |
556-96-7 | |
87°C | |
98% | |
C8H9Br | |
1.5475 to 1.5495 | |
1.36 | |
LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
1-bromo-3,5-dimethylbenzene | |
136357 | |
98% |
Chemical Identifiers
556-96-7 | |
185.06 | |
5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene | |
1-bromo-3,5-dimethylbenzene |
C8H9Br | |
LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
136357 | |
CC1=CC(=CC(=C1)Br)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doc
GHS Signal Word: Warning
RUO â Research Use Only