missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-m-xylene, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC294001000
Specifications
5-Bromo-m-xylene | |
87°C | |
Glass bottle | |
1.36 | |
100g | |
98% | |
(CH3)2C6H3Br | |
LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
1-bromo-3,5-dimethylbenzene | |
136357 | |
Liquid |
1.3600g/mL | |
Authentic | |
1.5475 to 1.5495 | |
Colorless to Yellow | |
556-96-7 | |
C8H9Br | |
1-bromo-3,5-dimethyl benzene, 1-bromo-3,5-dimethyl-benzene, 3,5-dimethylbromobenzene, 5-bromo-1,3-dimethylbenzene, 5-bromo-1,3-xylene, 5-bromo-3,5-dimethylbenzene, 5-bromo-m-xylene, 5-bromo-meta-xylene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo | |
CC1=CC(=CC(=C1)Br)C | |
185.06 | |
185.06 | |
98% |
Chemical Identifiers
556-96-7 | |
185.06 | |
1-bromo-3,5-dimethyl benzene, 1-bromo-3,5-dimethyl-benzene, 3,5-dimethylbromobenzene, 5-bromo-1,3-dimethylbenzene, 5-bromo-1,3-xylene, 5-bromo-3,5-dimethylbenzene, 5-bromo-m-xylene, 5-bromo-meta-xylene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo | |
1-bromo-3,5-dimethylbenzene |
C8H9Br | |
LMFRTSBQRLSJHC-UHFFFAOYSA-N | |
136357 | |
CC1=CC(=CC(=C1)Br)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doc
GHS Signal Word: Warning