missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-DL-tryptophan, 99%, ACROS Organics™

Chemical Identifiers

CAS 6548-09-0
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C11H11BrN2O2
Formula Weight 283.12
MDL Number MFCD00005648
Chemical Name or Material 5-Bromo-DL-tryptophan
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC107515000 SDS Fisher Scientific Acros
AC107515000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6548-09-0
100.0
C11H11BrN2O2
MFCD00005648
≥98.5%
98.5
99%
283.12
5-Bromo-DL-tryptophan

Specifications

6548-09-0
100.0
99%
C11H11BrN2O2
MFCD00005648
Off-white to light beige
500mg
≥98.5%
98.5
Fine Crystalline Powder
Authentic
283.12
Glass bottle
261° to 267°C (dec.)
5-Bromo-DL-tryptophan