Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-7-nitroindole, 99%

$94.93 - $268.20

Chemical Identifiers

Chemical Name or Material 5-Bromo-7-nitroindole
CAS 165669-16-9
Molecular Formula C8H5BrN2O2
Formula Weight 241.05
MDL Number MFCD00619260
Beilstein 8139934
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1492303

  SDS
alfa aesar™
L1492303
1g Each for $268.20

AAL14923MD

  SDS
alfa aesar™
L14923MD
250mg Each for $94.93
Description & Specifications

Specifications

Formula Weight 241.05
Molecular Formula C8H5BrN2O2
Health Hazard 1 Warning
Melting Point 211° to 214°C
Quantity 1g
Assay 99%
MDL Number MFCD00619260
CAS 165669-16-9
Beilstein 8139934
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Bromo-7-nitroindole

Applications
5-Bromo-7-nitroindole is used in gas chromatography analysis. It is also used as pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.