missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-7-azaindole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437430010

 View more versions of this product

Catalog No. AC437430010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Bromo-7-azaindole
Glass bottle
179.7°C to 180.7°C
183208-35-7
100.0
C7H5BrN2
LPTVWZSQAIDCEB-UHFFFAOYSA-N
5-bromo-1H-pyrrolo[2,3-b]pyridine
10307932
Crystalline Powder
Authentic
White to Yellow
1g
96.0
96% min. (HPLC)
1h-pyrrolo 2,3-b pyridine, 5-bromo, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo azaindole, 5-bromo,7-azaindole, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromo-1h-pyrrolo 2,3-b pyridine, 5-bromo-7-azaindole, 5-bromopyrrolo 2,3-b pyridine
C1=CNC2=NC=C(C=C21)Br
197.04
197.04
97%

Chemical Identifiers

183208-35-7
197.04
1h-pyrrolo 2,3-b pyridine, 5-bromo, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo azaindole, 5-bromo,7-azaindole, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromo-1h-pyrrolo 2,3-b pyridine, 5-bromo-7-azaindole, 5-bromopyrrolo 2,3-b pyridine
5-bromo-1H-pyrrolo[2,3-b]pyridine
C7H5BrN2
LPTVWZSQAIDCEB-UHFFFAOYSA-N
10307932
C1=CNC2=NC=C(C=C21)Br
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

SDS Documents