Please login to your online account to display your discounted pricing

5-Bromo-6-chloro-2-picoline, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Bromo-6-chloro-2-picoline
CAS 185017-72-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H5BrClN
Formula Weight 206.5
MDL Number MFCD03095218
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC360800050

  SDS
fisher scientific acros
AC360800050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC360800010

  SDS
thermo fisher scientific
AC360800010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 206.5
Physical Form Low Melting Solid or Liquid
Quantity 5g
Color Yellow
CAS Min % 98.0
CAS Max % 100.0
CAS 185017-72-5
Chemical Name or Material 5-Bromo-6-chloro-2-picoline
Molecular Formula C6H5BrClN
Assay Percent Range 98+%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03095218
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.