missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-methyl-2-phenyl-1,3-thiazole, ≥97%, Maybridge

$84.21 - $84.21

Chemical Identifiers

CAS 28771-82-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8BrNS
Formula Weight 254.15
MDL Number MFCD04974049
Synonym 5-Bromo-4-methyl-2-phenyl-1, 3-thiazole (IUPAC)
Chemical Name or Material 5-Bromo-4-methyl-2-phenyl-1, 3-thiazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC29210DA SDS Fisher Scientific Company
CC29210DA
Amber Glass Bottle 1g Each for $84.21
CC29210EA SDS Fisher Scientific Company
CC29210EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28771-82-6
100.0
C10H8BrNS
MFCD04974049
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole
96.5
97%
254.15
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole (IUPAC)
97%

Specifications

28771-82-6
100.0
1g
C10H8BrNS
MFCD04974049
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole
96.5
Amber Glass Bottle
97%
254.15
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole (IUPAC)
97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.