Please login to your online account to display your discounted pricing

5-Bromo-4-Indoyl-α-D-galactopyranoside, >99%, For TLC analysis, MP Biomedicals™


5-Bromo-4-chloro-3-indolyl-α-D-galactopyranoside is an α-galactoside substrate which differentiates α-galactosidase positive yeast strains or bacteria

Manufacturer: mp biomedicals  19392180

 View more versions of this product

Catalog No. ICN19392180

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-Indoyl-α-D-galactopyranoside
Name Note For TLC analysis
CAS 107021-38-5
Physical Form White powder
Assay Percent Range > 99%
Molecular Formula C14H15BrClNO6
Formula Weight 408.63g/mol
MDL Number MFCD00063780
Synonym 5-Bromo-4-chloro-3-indolyl- α-D -Galactoside

Specifications

Chemical Name or Material 5-Bromo-4-Indoyl-α-D-galactopyranoside
Name Note For TLC analysis
CAS 107021-38-5
Physical Form White powder
Assay Percent Range > 99%
Molecular Formula C14H15BrClNO6
Formula Weight 408.63g/mol
MDL Number MFCD00063780
Synonym 5-Bromo-4-chloro-3-indolyl- α-D -Galactoside