Please login to your online account to display your discounted pricing

5-Bromo-4-Indoyl-α-D-galactopyranoside, >99%, For TLC analysis, MP Biomedicals™

5-Bromo-4-chloro-3-indolyl-α-D-galactopyranoside is an α-galactoside substrate which differentiates α-galactosidase positive yeast strains or bacteria

$54.86 - $242.26

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-Indoyl-α-D-galactopyranoside
Name Note For TLC analysis
CAS 107021-38-5
Physical Form White powder
Assay Percent Range > 99%
Molecular Formula C14H15BrClNO6
Formula Weight 408.63g/mol
MDL Number MFCD00063780
Synonym 5-Bromo-4-chloro-3-indolyl- α-D -Galactoside
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19392180

 
mp biomedicals
19392180
Each for $242.26

ICN19392105

 
mp biomedicals
19392105
Each for $54.86
Description & Specifications

Specifications

Chemical Name or Material 5-Bromo-4-Indoyl-α-D-galactopyranoside
Name Note For TLC analysis
CAS 107021-38-5
Physical Form White powder
Assay Percent Range > 99%
Molecular Formula C14H15BrClNO6
Formula Weight 408.63g/mol
MDL Number MFCD00063780
Synonym 5-Bromo-4-chloro-3-indolyl- α-D -Galactoside