Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-4-fluoro-2-methylaniline, 96%

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-fluoro-2-methylaniline
CAS 627871-16-3
Formula Weight 204.04
MDL Number MFCD05865218
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6248003

 
alfa aesar™
H6248003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6248006

 
alfa aesar™
H6248006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 86° to 90°C
Formula Weight 204.04
Chemical Name or Material 5-Bromo-4-fluoro-2-methylaniline
Physical Form Solid
Assay 96%
Quantity 1g
MDL Number MFCD05865218
CAS 627871-16-3