Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-4-fluoro-1,3-benzodioxole, 96%

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-fluoro-1,3-benzodioxole
CAS 1226808-75-8
Formula Weight 219.01
MDL Number MFCD16295053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6294603

 
alfa aesar™
H6294603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62946MD

 
alfa aesar™
H62946MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 219.01
Chemical Name or Material 5-Bromo-4-fluoro-1,3-benzodioxole
Physical Form Solid
Assay 96%
Quantity 1g
MDL Number MFCD16295053
CAS 1226808-75-8