missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-chloroindoxyl Acetate 98.0+%, TCI America™

$81.86 - $241.03

Chemical Identifiers

CAS 3252-36-6
Molecular Formula C10H7BrClNO2
Molecular Weight (g/mol) 288.525
MDL Number MFCD00037932
InChI Key WPWLFFMSSOAORQ-UHFFFAOYSA-N
Synonym 5-Bromo-4-chloro-3-indolyl Acetate, 3-Acetoxy-4-chloro-5-bromoindole
PubChem CID 65116
IUPAC Name (5-bromo-4-chloro-1H-indol-3-yl) acetate
SMILES CC(=O)OC1=CNC2=C1C(=C(C=C2)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B483050MG
View Documents
Tci America
B483050MG
50mg Each for $81.86
 
B4830200MG
View Documents
Tci America
B4830200MG
200mg Each for $241.03
 
Specifications

Chemical Identifiers

3252-36-6
288.525
WPWLFFMSSOAORQ-UHFFFAOYSA-N
65116
CC(=O)OC1=CNC2=C1C(=C(C=C2)Br)Cl
C10H7BrClNO2
MFCD00037932
5-Bromo-4-chloro-3-indolyl Acetate, 3-Acetoxy-4-chloro-5-bromoindole
(5-bromo-4-chloro-1H-indol-3-yl) acetate

Specifications

White
50mg
C10H7BrClNO2
5-Bromo-4-chloro-3-indolyl Acetate, 3-Acetoxy-4-chloro-5-bromoindole
CC(=O)OC1=CNC2=C1C(=C(C=C2)Br)Cl
288.525
288.53
≥98.0% (HPLC)
107°C
3252-36-6
MFCD00037932
WPWLFFMSSOAORQ-UHFFFAOYSA-N
(5-bromo-4-chloro-1H-indol-3-yl) acetate
65116
Crystal-Powder at 20°C
5-Bromo-4-chloroindoxyl Acetate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator