Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-4-chloroindoxyl 1,3-diacetate, 99%

$1,670.12 - $1,670.12

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-chloroindoxyl 1,3-diacetate
CAS 3030-06-6
Molecular Formula C12H9BrClNO3
Formula Weight 330.57
MDL Number MFCD00040632
Synonym 1-Acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4336709

  SDS
alfa aesar™
4336709
10g Each for $1,670.12
Description & Specifications

Specifications

Formula Weight 330.57
Chemical Name or Material 5-Bromo-4-chloroindoxyl 1,3-diacetate
Physical Form Powder
Assay 99%
Molecular Formula C12H9BrClNO3
MDL Number MFCD00040632
CAS 3030-06-6
Melting Point 165° to 166°C
Synonym 1-Acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate
Quantity 10g