Please login to your online account to display your discounted pricing

5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt, 98%, Molecular biology reagent grade, For TLC analysis, MP Biomedicals™

5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt is β-glucuronidase substrate which forms an intense blue precipitate upon enzymatic action

$180.95 - $903.58

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt
Name Note For TLC analysis
CAS 129541-41-9
Purity Grade Molecular Biology Reagent Grade
Physical Form Powder
Molecular Formula C14H13BrClNO7Na
Formula Weight 444.6
MDL Number MFCD00135782
Synonym BC-Indicator
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19481310

 
mp biomedicals
19481310
10mg Each for $180.95

ICN19481325

 
mp biomedicals
19481325
25mg Each for $296.36

ICN19481380

 
mp biomedicals
19481380
100mg Each for $903.58
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 444.6
Purity Grade Molecular Biology Reagent Grade
Color White
Quantity 10mg
Synonym BC-Indicator
MDL Number MFCD00135782
CAS 129541-41-9
Name Note For TLC analysis
Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt
Molecular Formula C14H13BrClNO7Na