Please login to your online account to display your discounted pricing

5-Bromo-4-chloro-3-indolyl sulfate potassium salt, 99%, ACROS Organics™

$298.25 - $298.25

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-chloro-3-indolyl sulfate potassium salt
Name Note 99%
CAS 6578-07-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H4BrClKNO4S
Formula Weight 364.63
MDL Number MFCD00056929
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC337232000

  SDS
thermo fisher scientific
AC337232000
Glass bottle 200mg Each for $298.25
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 364.63
Quantity 200mg
Color Almost white to light yellow
CAS Max % 100.0
CAS 6578-07-0
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl sulfate potassium salt
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H4BrClKNO4S
MDL Number MFCD00056929
Synonym X-Sulfate potassium salt
CAS Min % 98.5
Infrared Spectrum Authentic