missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-chloro-3-indolyl Phosphate p-Toluidine Salt [for Biochemical Research], TCI America™

$88.61 - $516.41

Chemical Identifiers

CAS 6578-06-9
Molecular Formula C15H15BrClN2O4P
Molecular Weight (g/mol) 433.62
MDL Number MFCD00040636
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N
Synonym X-Phosphate p-Toluidine Salt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12391G View Documents TCI America
B12391G
1g Each for $516.41
 
B1239100MG View Documents TCI America
B1239100MG
100mg Each for $88.61
 
Specifications

Chemical Identifiers

6578-06-9
433.62
QEIFSLUFHRCVQL-UHFFFAOYSA-N
C15H15BrClN2O4P
MFCD00040636
X-Phosphate p-Toluidine Salt

Specifications

Undesignated
6578-06-9
MFCD00040636
X-Phosphate p-Toluidine Salt
433.62
Crystal-Powder at 20°C
5-Bromo-4-chloro-3-indolyl Phosphate p-Toluidine Salt [for Biochemical Research]
1g
C15H15BrClN2O4P
2811
QEIFSLUFHRCVQL-UHFFFAOYSA-N
433.62
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer