missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98+%

Manufacturer:  Alfa Aesar™ H5626506

 View more versions of this product

Catalog No. AAH5626506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
It is an histochemical substrate for alkaline phosphatase. For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

Solubility
Soluble in water and toluene.

Notes
Moisture and light sensitive. Store in dry place. Keep away from oxidizing agents, heat and light.
Specifications

Specifications

5-Bromo-4-chloro-3-indolyl phosphate disodium salt
102185-33-1
MFCD00149370
sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip®, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
C1=CC(=C(C2=C1NC=C2OP(=O)([O-])[O-])Cl)Br.[Na+].[Na+]
370.43
370.43
5g
C8H4BrClNNa2O4P
4722243
OAZUOCJOEUNDEK-UHFFFAOYSA-L
disodium;(5-bromo-4-chloro-1H-indol-3-yl) phosphate
6097197
≥98%

Chemical Identifiers

102185-33-1
370.43
OAZUOCJOEUNDEK-UHFFFAOYSA-L
6097197
C1=CC(=C(C2=C1NC=C2OP(=O)([O-])[O-])Cl)Br.[Na+].[Na+]
C8H4BrClNNa2O4P
MFCD00149370
sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip®, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
disodium;(5-bromo-4-chloro-1H-indol-3-yl) phosphate
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C

Documents