Please login to your online account to display your discounted pricing

X-Gal, 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, Fisher BioReagents™


For blue/white colony screening, immunoblotting and immunocytochemical assays

Quantity 1g
Packaging Glass Bottle

Manufacturer: fisher scientific company  BP16151

 View more versions of this product

Catalog No. BP1615-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 408.63
Melting Point 230°C
Quantity 1g
Molecular Formula C14H15BrClNO6
Alpha Vector Bromo-4-chloro-3-indolyl-[beta]-D-galactopyranoside (X-Gal)
Color White
Merck Index 15, 10272
Packaging Glass Bottle
Specific Rotation -56 to -64° (+ or -)
CAS 7240-90-6
ChemAlert Storage Symbol Gray
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl- β-D-galactopyranoside
Molar Absorption Coefficient 4000 /g-cm. min.
TLC Single spot
Synonym X-Gal
Identification Pass Test
Recommended Storage -20°C

Specifications

Chemical Name or Material 5-Bromo-4-chloro-3-indolyl- β-D-galactopyranoside
CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Formula Weight 408.63
Synonym X-Gal

  • Chromogenic substrate used for blue-white selection of recombinant bacterial colonies with the lac+ genotype
  • Forms blue precipitate when hydrolyzed by β-galactosidase
  • Soluble in DMF and Methanol

Accessories
 

-20°C

SDS & Safety Documents