missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%, Thermo Scientific Chemicals

Substrate for beta-glucuronidase

Supplier:  Thermo Scientific Chemicals J6436003

 View more versions of this product

Catalog No. AAJ6436003



Description

Description

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

Solubility
Soluble in water at 10mg/ml

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt
White
MFCD00135782
Light sensitive
Soluble in water at 10mg/ml
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]
444.594
444.59
Powder
129541-41-9
C14H12BrClNNaO7
1 g
x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
IBLSVGDGSKUDCT-ILIJQVQCSA-M
sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
53384407
98%

Chemical Identifiers

129541-41-9
444.594
IBLSVGDGSKUDCT-ILIJQVQCSA-M
53384407
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]
C14H12BrClNNaO7
MFCD00135782
x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only