Please login to your online account to display your discounted pricing

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%, ACROS Organics™


Quantity 25mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC331190250

 View more versions of this product

Catalog No. AC331190250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 521.79
Physical Form Powder, Needles or Crystals
Quantity 25mg
Color White to off-white or colorless
TSCA TSCA
CAS Max % 100.0
Health Hazard 2 MOISTURE SENSITIVE
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate
Health Hazard 1 LIGHT SENSITIVE
CAS 114162-64-0
Molecular Formula C6H13N·xH2O
CAS Min % 97.5
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00058543
Infrared Spectrum Authentic
Synonym X-Gluc
Packaging Glass bottle
Water 5% max. (K.F.)

Specifications

Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate
Name Note 98%
CAS 114162-64-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H13N·xH2O
Formula Weight 521.79
MDL Number MFCD00058543
Synonym X-Gluc
Accessories

LIGHT SENSITIVE

MOISTURE SENSITIVE

SDS & Safety Documents