missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl beta-D-glucoside, 99%, Thermo Scientific Chemicals
Substrate for beta→-D-glucosidase
Supplier: Thermo Scientific Chemicals J6567703
Description
Substrate for beta--D-glucosidase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Bromo-4-chloro-3-indolyl beta-D-glucoside | |
White | |
MFCD00063690 | |
1552603 | |
Soluble in DMF or DMSO | |
OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O | |
408.63 | |
CHEBI:75424 | |
99% |
15548-60-4 | |
C14H15BrClNO6 | |
1 g | |
x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside | |
OPIFSICVWOWJMJ-LNNRFACYSA-N | |
(2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
84982 | |
408.63 | |
Crystalline Powder |
Chemical Identifiers
15548-60-4 | |
408.63 | |
OPIFSICVWOWJMJ-LNNRFACYSA-N | |
84982 | |
(2S,3R,4S,5S,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
C14H15BrClNO6 | |
MFCD00063690 | |
x-glc, x-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucoside, 5-bromo-4-chloro-3-indolyl-beta-d-glucopyranoside, 5-bromo-4-chloroindol-3-yl-beta-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl b-d-glucopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-glucoside, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, 2s,3r,4s,5s,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, indoxyl-glucoside | |
CHEBI:75424 | |
OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)[C@H](O)[C@@H](O)[C@@H]1O |
Safety and Handling
EINECSNumber : 239-603-0
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only