missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 227771000

 View more versions of this product

Catalog No. AC227771000



Specifications

Specifications

5-Bromo-4-chloro-3-indolyl-β-D-galactoside
Glass bottle
1% max. (K.F.)
230.0°C
7240-90-6
100.0
C14H15BrClNO6
15,10272
OPIFSICVWOWJMJ-AEOCFKNESA-N
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
65181
408.63
99%
Authentic
− 64.00
White
100mg
98.5
98.5% min. (HPLC)
MFCD00005666
5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, indoxyl-gal, unii-v595og374w, x-gal
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
408.63
CHEBI:75055
Powder
Pure

Chemical Identifiers

7240-90-6
408.63
OPIFSICVWOWJMJ-AEOCFKNESA-N
65181
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C14H15BrClNO6
MFCD00005666
5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, indoxyl-gal, unii-v595og374w, x-gal
CHEBI:75055
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
Safety and Handling

Safety and Handling

EINECSNumber : 230-640-8

SDS Documents