missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 227770100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Bromo-4-chloro-3-indolyl-β-D-galactoside | |
98.5 | |
230.0°C | |
Authentic | |
Glass bottle | |
MFCD00005666 | |
15,10272 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br | |
408.63 | |
CHEBI:75055 | |
99% | |
Powder |
7240-90-6 | |
100.0 | |
White | |
98.5% min. (HPLC) | |
C14H15BrClNO6 | |
10 mg | |
− 64.00 | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
65181 | |
408.63 | |
Pure |
Chemical Identifiers
7240-90-6 | |
408.63 | |
OPIFSICVWOWJMJ-AEOCFKNESA-N | |
65181 | |
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
C14H15BrClNO6 | |
MFCD00005666 | |
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal | |
CHEBI:75055 | |
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br |
Safety and Handling
EINECSNumber : 230-640-8
RUO – Research Use Only