Please login to your online account to display your discounted pricing

5-Bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one
CAS 300574-36-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5BrN2O2
Formula Weight 217.02
MDL Number MFCD00168437
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RJC02150CB

  SDS
fisher scientific company
RJC02150CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.02
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-Bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one
Assay Percent Range 97%
CAS 300574-36-1
MDL Number MFCD00168437
Packaging Amber Glass Bottle
Molecular Formula C6H5BrN2O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.