Please login to your online account to display your discounted pricing

(5-Bromo-3-pyridinyl)methylamine, 97%, Maybridge

$231.16 - $231.16

Chemical Identifiers

Chemical Name or Material (5-Bromo-3-pyridinyl)methylamine
CAS 135124-70-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7BrN2
Formula Weight 187.04
MDL Number MFCD06212876
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC67113DA

 
fisher scientific company
CC67113DA
Amber Glass Bottle 1g Each for $231.16

CC67113DE

 
fisher scientific company
CC67113DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.04
Quantity 1g
Quantity 5Pack
Type Spherical Joint
Assay 97%
CAS Max % 100.0
Chemical Name or Material (5-Bromo-3-pyridinyl)methylamine
Assay Percent Range 97%
Molecular Formula C6H7BrN2
CAS 135124-70-8
MDL Number MFCD06212876
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.