missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-3-pyridinecarbonitrile, TCI America™

$67.22 - $198.61

Chemical Identifiers

CAS 35590-37-5
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00174363
InChI Key FTFFHWWIPOQCBC-UHFFFAOYSA-N
Synonym 5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
PubChem CID 736793
IUPAC Name 5-bromopyridine-3-carbonitrile
SMILES C1=C(C=NC=C1Br)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37131G View Documents TCI America
B37131G
1g Each for $67.22
 
B37135G View Documents TCI America
B37135G
5g Each for $198.61
 
Specifications

Chemical Identifiers

35590-37-5
183.008
FTFFHWWIPOQCBC-UHFFFAOYSA-N
736793
C1=C(C=NC=C1Br)C#N
C6H3BrN2
MFCD00174363
5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
5-bromopyridine-3-carbonitrile

Specifications

White-Yellow
1g
C6H3BrN2
3439
FTFFHWWIPOQCBC-UHFFFAOYSA-N
5-bromopyridine-3-carbonitrile
736793
Crystal-Powder at 20°C
5-Bromo-3-pyridinecarbonitrile
105°C
35590-37-5
MFCD00174363
5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
C1=C(C=NC=C1Br)C#N
183.008
183.01
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No