Learn More
5-Bromo-3-pyridineacetic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1357906
Description
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
5-Bromo-3-pyridineacetic acid | |
182°C to 185°C | |
MFCD00829308 | |
388264 | |
Soluble in water. | |
C1=C(C=NC=C1Br)CC(=O)O | |
216.034 | |
216.03 |
39891-12-8 | |
C7H6BrNO2 | |
5 g | |
5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid | |
UETIDNDXXGCJCE-UHFFFAOYSA-N | |
2-(5-bromopyridin-3-yl)acetic acid | |
2802539 | |
≥98% |
Chemical Identifiers
39891-12-8 | |
216.034 | |
UETIDNDXXGCJCE-UHFFFAOYSA-N | |
2802539 | |
C1=C(C=NC=C1Br)CC(=O)O |
C7H6BrNO2 | |
MFCD00829308 | |
5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid | |
2-(5-bromopyridin-3-yl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only