missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-methylpyridine-2-carboxylic acid, 97%

$39.78 - $467.26

Chemical Identifiers

CAS 886365-43-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD07375125
InChI Key GSOCTKHBFREQCV-UHFFFAOYSA-N
Synonym 5-bromo-2-carboxy-3-methylpyridine, 5-bromo-3-methylpicolinic acid, 5-bromo-3-methyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-methyl, ksc447q6l, 5-bromanyl-3-methyl-pyridine-2-carboxylic acid, 5-bromo-2-carboxy-3-methylpyridine
PubChem CID 45789779
IUPAC Name 5-bromo-3-methylpyridine-2-carboxylic acid
SMILES CC1=CC(=CN=C1C(=O)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64220MD View Documents Alfa Aesar™
H64220MD
250mg Each for $39.78
 
AAH6422003 View Documents Alfa Aesar™
H6422003
1g Each for $117.66
 
AAH6422006 View Documents Alfa Aesar™
H6422006
5g Each for $467.26
 
Specifications

Chemical Identifiers

886365-43-1
216.034
GSOCTKHBFREQCV-UHFFFAOYSA-N
45789779
CC1=CC(=CN=C1C(=O)O)Br
C7H6BrNO2
MFCD07375125
5-bromo-2-carboxy-3-methylpyridine, 5-bromo-3-methylpicolinic acid, 5-bromo-3-methyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-methyl, ksc447q6l, 5-bromanyl-3-methyl-pyridine-2-carboxylic acid, 5-bromo-2-carboxy-3-methylpyridine
5-bromo-3-methylpyridine-2-carboxylic acid

Specifications

White
886365-43-1
MFCD07375125
GSOCTKHBFREQCV-UHFFFAOYSA-N
5-bromo-3-methylpyridine-2-carboxylic acid
45789779
Powder
5-Bromo-3-methylpyridine-2-carboxylic acid
250mg
C7H6BrNO2
5-bromo-2-carboxy-3-methylpyridine, 5-bromo-3-methylpicolinic acid, 5-bromo-3-methyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo-3-methyl, ksc447q6l, 5-bromanyl-3-methyl-pyridine-2-carboxylic acid, 5-bromo-2-carboxy-3-methylpyridine
CC1=CC(=CN=C1C(=O)O)Br
216.034
216.03
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning