Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine, 95%

$183.18 - $183.18

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
CAS 245765-92-8
Molecular Formula C12H17BrN2O
Formula Weight 285.18
MDL Number MFCD09037448
Synonym N-(5-Bromo-3-methyl-2-pyridyl)-N-methylpivalamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27468MD

 
alfa aesar™
H27468MD
250mg Each for $183.18
Description & Specifications

Specifications

Formula Weight 285.18
Chemical Name or Material 5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
Assay 95%
Molecular Formula C12H17BrN2O
MDL Number MFCD09037448
CAS 245765-92-8
Synonym N-(5-Bromo-3-methyl-2-pyridyl)-N-methylpivalamide
Quantity 250mg
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.