missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-methyl-2-(methylamino)pyridine, 96%

Manufacturer:  Alfa Aesar™ H54360MD

 View more versions of this product

Catalog No. AAH54360MD



Specifications

Specifications

5-Bromo-3-methyl-2-(methylamino)pyridine
250mg
C7H9BrN2
2-pyridinamine, 5-bromo-n,3-dimethyl
CC1=CC(=CN=C1NC)Br
201.067
201.06
62°C to 64°C
245765-66-6
MFCD10000482
QTBNIBCNVQMGPP-UHFFFAOYSA-N
5-bromo-N,3-dimethylpyridin-2-amine
18329367
96%

Chemical Identifiers

245765-66-6
201.067
QTBNIBCNVQMGPP-UHFFFAOYSA-N
18329367
CC1=CC(=CN=C1NC)Br
C7H9BrN2
MFCD10000482
2-pyridinamine, 5-bromo-n,3-dimethyl
5-bromo-N,3-dimethylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

Documents