Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine, 95%

$93.27 - $209.34

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine
CAS 446299-84-9
Molecular Formula C11H15BrN2O
Formula Weight 271.16
MDL Number MFCD09037447
Synonym N-(5-Bromo-3-methyl-2-pyridyl)pivalamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2731703

 
alfa aesar™
H2731703
1g Each for $209.34

AAH27317MD

 
alfa aesar™
H27317MD
250mg Each for $93.27
Description & Specifications

Specifications

Formula Weight 271.16
Chemical Name or Material 5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine
Assay 95%
Molecular Formula C11H15BrN2O
MDL Number MFCD09037447
CAS 446299-84-9
Melting Point 125° to 128°C
Synonym N-(5-Bromo-3-methyl-2-pyridyl)pivalamide
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.