Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-iodo-7-azaindole, 97%

$63.59 - $204.92

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-iodo-7-azaindole
CAS 757978-18-0
Molecular Formula C7H4BrIN2
Formula Weight 322.93
MDL Number MFCD07779519
Synonym 5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5004103

  SDS
alfa aesar™
H5004103
1g Each for $204.92

AAH50041MD

  SDS
alfa aesar™
H50041MD
250mg Each for $63.59
Description & Specifications

Specifications

Formula Weight 322.93
Synonym 5-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Chemical Name or Material 5-Bromo-3-iodo-7-azaindole
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H4BrIN2
CAS 757978-18-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD07779519
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.