Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-iodo-2-methoxypyridine, 97%

$242.59 - $1,074.32

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-iodo-2-methoxypyridine
CAS 578007-66-6
Molecular Formula C6H5BrINO
Formula Weight 313.92
MDL Number MFCD05663522
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5006103

  SDS
alfa aesar™
H5006103
1g Each for $242.59

AAH5006106

  SDS
alfa aesar™
H5006106
5g Each for $1,074.32
Description & Specifications

Specifications

Formula Weight 313.92
Chemical Name or Material 5-Bromo-3-iodo-2-methoxypyridine
Assay 97%
Molecular Formula C6H5BrINO
MDL Number MFCD05663522
CAS 578007-66-6
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
5-Bromo-3-iodo-2-methoxypyridine is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.