Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-hydroxyoxindole, 96%

$80.81 - $321.73

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-hydroxyoxindole
CAS 99304-37-7
Molecular Formula C8H6BrNO2
Formula Weight 228.05g/mol
MDL Number MFCD00052491
Synonym 5-Bromo-3-hydroxy-2-indolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1586906

 
alfa aesar™
A15869-06
5g Each for $80.81

AAA1586914

 
alfa aesar™
A15869-14
25g Each for $321.73
Description & Specifications

Specifications

Melting Point ≈205°C (dec.)
Quantity 5g
Chemical Name or Material 5-Bromo-3-hydroxyoxindole
CAS 99304-37-7
Assay 96%
Molecular Formula C8H6BrNO2
Formula Weight 228.05g/mol
MDL Number MFCD00052491
Synonym 5-Bromo-3-hydroxy-2-indolinone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.