missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452570010

 View more versions of this product

Catalog No. AC452570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
68°C to 70°C (25.0 Torr)
1262412-30-5
C6H2BrF4N
ZYPJKTSNQGVERF-UHFFFAOYSA-N
5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
54772300
97%
Glass Bottle
1g
96% min. (GC)
5-bromo-3-fluoro-2-trifluoromethyl pyridine
C1=C(C(=NC=C1Br)C(F)(F)F)F
243.98
243.98

Chemical Identifiers

1262412-30-5
243.98
5-bromo-3-fluoro-2-trifluoromethyl pyridine
5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
C6H2BrF4N
ZYPJKTSNQGVERF-UHFFFAOYSA-N
54772300
C1=C(C(=NC=C1Br)C(F)(F)F)F
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents